Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Mouse Surgery Swivel Platform
SDP

Catalog No. 01259173
Change view
Click to view available options
Dimensions (L x W x H):
11.5 x 7 x 6 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
01-259-173 11.5 x 7 x 6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
N/A
Catalog No. 01-259-173 Supplier Poltex Supplier No. MOUSWIV
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
N/A

Mouse surgery or procedure platform with an easily adjustable swivel arm. Durable and cleanable PETG construction.

  • Mouse surgery or procedure platform
  • Features an easily adjustable swivel arm
  • Durable and cleanable PETG construction
  • Consumables not included

Specifications

Autoclavable Not Autoclavable
Capacity (English) One Mouse
Color Clear
Construction PETG
Animal Mouse
For Use With (Application) Mouse Surgery or Procedure Platform
Disposable Reusable
Includes Adjustable Swivel Arm
Material PETG
Sterility Non-sterile
Sterilization Method Cleaning Solution, Mild Soap or Detergent
Dimensions (L x W x H) 11.5 x 7 x 6 in.
Weight Range 1 lb.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.