Learn More
Biomatik Corporation Mouse Synapsin II (SYN2) ELISA Kit
Supplier:  Biomatik Corporation EKU11452
Encompass_Preferred
Mouse Synapsin II (SYN2) ELISA Kit for detecting SYN2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 30pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q64332. Gene ID: 20965. Target Synonyms: SYNII; SYNIIa; SYNIIb. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8142