Learn More
BIOHIPPO Mouse SYP Microsample
SDP
Catalog No. 5000315677
Encompass
Mouse SYP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SYP in serum plasma tissue homogenates cerebrospinal fluid and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 10 79 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse SYP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse SYP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse SYP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15677 Supplier BIOHIPPO Supplier No. BHE15212482
Mouse SYP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of SYP in serum plasma tissue homogenates cerebrospinal fluid and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 10 79 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse SYP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse SYP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse SYP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color