Learn More
American Research Products Inc Mouse T lymphocyte actiVation
Catalog No. 5000327730
Encompass
Mouse T-lymphocyte actiVation antigen CD80 (CD80) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27730 Supplier American Research Products Inc Supplier No. CSBEL004959MO
Mouse T-lymphocyte actiVation antigen CD80 (CD80) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich