Learn More
Sino Biological Anti-TROP-2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50922RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse TROP-2 / TACSTD2 (rM TROP-2 / TACSTD2; Catalog#50922-M08H; Q8BGV3; Met1-Gln270). TROP-2 / TACSTD2 specific IgG was purified by Mouse TROP-2 / TACSTD2 affinity chromatography.
Catalog No. 50-241-2457