Learn More
TRIBIOSCIENCE INC Mouse Tail Fast DNA Extraction Kit, 1000 Samples
SDP
Catalog No. NC2607291
Encompass
Mouse tail DNA extraction kit provides a simple and rapid method to isolate the total DNA from mouse tail for genotyping. No organic extraction and alcohol precipitation are needed, and multiple samples can be easily processed simultaneously. The extraction time is only 10 mins.
Catalog No. NC2607291 Supplier TRIBIOSCIENCE INC Supplier No. TBS60051000
Mouse tail DNA extraction kit provides a simple and rapid method to isolate the total DNA from mouse tail for genotyping. No organic extraction and alcohol precipitation are needed, and multiple samples can be easily processed simultaneously. The extraction time is only 10 mins.