Learn More
DAN KAR CORP MOUSE TAIL ILLUMINATOR
Catalog No. NC1713961
Encompass_Preferred
The MTI efficiently illuminates and warms a mouse tail for easy injection or bleeding. Each unit comes with a restraining tube and illumination source. Extra tubes can be purchased at nominal cost.
Catalog No. NC1713961 Supplier DAN KAR CORP Supplier No. MTI
The MTI efficiently illuminates and warms a mouse tail for easy injection or bleeding. Each unit comes with a restraining tube and illumination source. Extra tubes can be purchased at nominal cost.