Description
- Specialized head cone securely immobilizes the mouse’s head and body
- Open cone design encourages natural, head-first positioning
- Avoids pinching or obstructing the animal’s nose
- Rectangular body prevents rolling during injection procedures
- Can be placed on either side to position the lateral tail vein upward
- Sliding, removable trap door simplifies animal loading and unloading
- Removable head cone provides additional handling flexibility
- Spacious chamber accommodates animals with flank tumors
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Animal | All Animals |
| Type | Restrainer |
| Disposable | Reusable |
| Size | One Size |
| Sterility | Non-sterile |