Learn More
American Research Products Inc Mouse tartrate resistant acid
Catalog No. 5000330214
Encompass
Mouse tartrate-resistant acid phosphatase 5b (TRACP-5b) ELISA Kit from CUSABIO Detection Range 0 078 mIU/mL-5 mIU/mL Sensitivity 0 02 mIU/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30214 Supplier American Research Products Inc Supplier No. CSBE08492M
Mouse tartrate-resistant acid phosphatase 5b (TRACP-5b) ELISA Kit from CUSABIO Detection Range 0 078 mIU/mL-5 mIU/mL Sensitivity 0 02 mIU/mL Sample serum plasma tissue homogenates Method Sandwich