Learn More
Assaypro MOUSE TAT COMPLEX ELISA KIT
Supplier:  Assaypro EMT10201
Encompass
The AssayMax Mouse TAT Complex ELISA kit is designed for detection of mouse TAT complex in plasma and cell culture samples. Assay employs a quantitative sandwich enzyme immunoassay technique that measures mouse TAT complex in less than 5 hours.
Catalog No. NC0707068