Learn More
Biomatik Corporation Mouse Taxilin Alpha (TXLNa) ELISA Kit
Supplier:  Biomatik Corporation EKU11460
Encompass_Preferred
Mouse Taxilin Alpha (TXLNa) ELISA Kit for detecting TXLNa in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6PAM1. Gene ID: 109658. Target Synonyms: IL14; TXLN; HMW-BCGF; High Molecular Weight B-Cell Growth Factor; Interleukin 14. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4862