Learn More
Raybiotech Inc MOUSE TCK-1 ELISA
Supplier:  Raybiotech Inc ELMTCK11
Encompass_Preferred
RayBio Mouse TCK-1 (CXCL7) ELISA Kit for cell culture supernatants, plasma, and serum samples. Pre-coated 96-well microplate kit with additional reagents. Detection Range: 14 pg/ml - 8000 pg/ml
Catalog No. 50-104-4797