Learn More
Sino Biological Recombinant Anti-Transcobalamin II Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50693R049100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TCN2 (rM TCN2; Catalog#50693-M08H; O88968; Met1-Trp430).
Catalog No. 50-255-3153