Learn More
Sino Biological Anti-Transcobalamin II Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50693RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse TCN2 (rM TCN2; Catalog#50693-M08H; O88968; Met 1-Trp 430). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3152