Learn More
Sino Biological Anti-Transcobalamin II Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50693RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse TCN2 (rM TCN2; Catalog#50693-M08H; O88968; Met 1-Trp 430). TCN2 specific IgG was purified by Mouse TCN2 affinity chromatography.
Catalog No. 50-255-3151