Learn More
Sino Biological Anti-TIE-2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51087T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Tie2 / CD202b / TEK (rM Tie2 / CD202b / TEK; Catalog#51087-M20B1; Q02858; Gln770-Ala1122). Tie2 / CD202b / TEK specific IgG was purified by Mouse Tie2 / CD202b / TEK affinity chromatography.
Catalog No. 50-241-2335