Learn More
American Research Products Inc Mouse terminal complement comp
Catalog No. 5000329506
Encompass
Mouse terminal complement compleX C5b-9 TCC C5b-9 ELISA Kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-29506 Supplier American Research Products Inc Supplier No. CSBE08710M
Mouse terminal complement compleX C5b-9 TCC C5b-9 ELISA Kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich