Learn More
Crystal Chem Inc Mouse Testosterone ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80552
Encompass
The Mouse Testosterone ELISA is used to quantify levels of mouse testosterone in serum and plasma using only a 10 µL sample volume. Highlights: Kit uses sample size of only 10 µL. Dynamic range of 0.1 - 25 ng/mL. No extraction needed, ready to use reagents. Highly precise with low CV's. High sensitivity of 0.066 ng/mL. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7726