Learn More
American Research Products Inc Mouse Testosterone T ELISA Ki
Catalog No. 5000326332
Encompass
Mouse Testosterone T ELISA Kit from CUSABIO Detection Range 15 6 pg/ml-1000 pg/ml Sensitivity 3 9 pg/ml Sample serum plasma Method Competitive
Catalog No. 50-003-26332 Supplier American Research Products Inc Supplier No. CSBE05101M
Mouse Testosterone T ELISA Kit from CUSABIO Detection Range 15 6 pg/ml-1000 pg/ml Sensitivity 3 9 pg/ml Sample serum plasma Method Competitive