Learn More
American Research Products Inc Mouse Tetranectin CLEC3B ELI
Catalog No. 5000328114
Encompass
Mouse Tetranectin (CLEC3B) ELISA kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28114 Supplier American Research Products Inc Supplier No. CSBEL005531MO
Mouse Tetranectin (CLEC3B) ELISA kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich