Learn More
BIOHIPPO Mouse TF Microsample
SDP
Catalog No. 5000314008
Encompass
Mouse TF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TF in Plasma It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse TF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse TF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse TF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14008 Supplier BIOHIPPO Supplier No. BHE15211980
Mouse TF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TF in Plasma It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse TF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse TF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse TF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color