Learn More
Sino Biological Anti-Transferrin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50817RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Transferrin / TF (Catalog#50817-M08H; Q921I1; Met1-His697). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8532