Learn More
Sino Biological Recombinant Anti-TFPI Antibody (HRP), Rabbit Monoclonal
Supplier:  Sino Biological 50131R103H100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse TFPI (rM TFPI ; Catalog#50131-M08H; NP_035706.1; Met1-Lys289), and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-255-3440