Learn More
Sino Biological Mouse TFPI2 Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785261
Encompass_Preferred
The Mouse TFPI2 ELISA Pair Set is for the quantitative determination of Mouse TFPI2.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse TFPI2 coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse TFPI2 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Mouse TFPI2 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse TFPI2 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85261 Supplier Sino Biological Supplier No. SEK510025
The Mouse TFPI2 ELISA Pair Set is for the quantitative determination of Mouse TFPI2.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Mouse TFPI2 coated on a 96-well plate. Standards and samples are added to the wells, and any Mouse TFPI2 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Mouse TFPI2 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Mouse TFPI2 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.