Learn More
Sino Biological Anti-Transferrin Receptor/TFR1/CD71 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50741T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Transferrin Receptor / TFRC / CD71 (Catalog#50741-M07H; Q62351; Cys89-Phe763). Transferrin Receptor / TFRC / CD71 specific IgG was purified by Mouse Transferrin Receptor / TFRC / CD71 affinity chromatography.
Catalog No. 50-240-9518