Learn More
Sino Biological Recombinant Mouse Transferrin Receptor/TFR1/CD71 Protein (ECD, His Tag)
Supplier:  Sino Biological 50741M07H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse TFRC (Q62351) (Cys 89-Phe 763) was fused with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-3111