Learn More
ACROBiosystems Mouse TGF-Beta 1 / TGFB1 Protein
Catalog No. 502055050
Encompass_Preferred
Recombinant Protein;50UG;Mouse TGFB1 Protein (TG1-M5218) is expressed from human 293 cells (HEK293). It contains AA Ala 279 - Ser 390 (Accession # P04202-1).;This protein carries no "tag". The protein has a calculated MW of 14.7 kDa.
Catalog No. 50-205-5050 Supplier ACROBiosystems Supplier No. TG1M521850UG
Recombinant Protein;50UG;Mouse TGFB1 Protein (TG1-M5218) is expressed from human 293 cells (HEK293). It contains AA Ala 279 - Ser 390 (Accession # P04202-1).;This protein carries no "tag". The protein has a calculated MW of 14.7 kDa.