Learn More
BIOHIPPO Mouse TGFb1 Microsample
SDP
Catalog No. 5000313690
Encompass
Mouse TGFb1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TGFb1 in serum platelet-poor plasma tissue homogenates cell culture supernates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 4 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse TGFb1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse TGFb1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse TGFb1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13690 Supplier BIOHIPPO Supplier No. BHE15212291
Mouse TGFb1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TGFb1 in serum platelet-poor plasma tissue homogenates cell culture supernates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 4 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse TGFb1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse TGFb1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse TGFb1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color