Learn More
Sino Biological Anti-TGF beta 2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50153RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse TGF-beta 2 (rM TGF-beta 2; Catalog#50153-M08H; NP_033393.2; Met1-Ser414). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3399