Learn More
Sino Biological Recombinant Mouse TGF beta 2 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 50153M08HB100
Encompass_Preferred
A DNA sequence encoding the mouse Tgfb2 (NP_033393.2) (Met1-Ser414) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-3401