Learn More
BIOHIPPO Mouse TGFbI Microsample
SDP
Catalog No. 5000314165
Encompass
Mouse TGFbI Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TGFbI in Serum plasma and other biological fluids It provides a detection range of 0 63-40 ng/mL with a sensitivity of 0 21 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse TGFbI Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse TGFbI Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse TGFbI biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14165 Supplier BIOHIPPO Supplier No. BHE15212587
Mouse TGFbI Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TGFbI in Serum plasma and other biological fluids It provides a detection range of 0 63-40 ng/mL with a sensitivity of 0 21 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse TGFbI Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse TGFbI Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse TGFbI biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color