Learn More
BIOHIPPO Mouse TGFbR2 Microsample
SDP
Catalog No. 5000315472
Encompass
Mouse TGFbR2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TGFbR2 in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 112 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse TGFbR2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse TGFbR2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse TGFbR2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15472 Supplier BIOHIPPO Supplier No. BHE15210976
Mouse TGFbR2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of TGFbR2 in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 112 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse TGFbR2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse TGFbR2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse TGFbR2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color