Learn More
Sino Biological Anti-TGF-beta RIII/TGFBR3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50542RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse TGFBR3 (rM TGFBR3; Catalog#50542-M08H; NP_035708.2; Met 1-Arg 399). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3220