Learn More
R&D Systems™ Mouse Th1 Cell Multi-Color Flow Cytometry Kit
Catalog No. FMC010
Change view
Click to view available options
:
25 Tests
Catalog No.
FMC010 25 Tests
1 options
This item is not returnable. View return policy
Catalog No. FMC010 Supplier R&D Systems™ Supplier No. FMC010
This item is not returnable. View return policy
For flow cytometric analysis of human Th1 cells using four fluorochrome-conjugated antibodies
Includes conjugated antibodies (and corresponding isotype controls): IFN-gamma-CFS (Clone 37895; rat IgG2A), IL-12 Rbeta2-APC (Clone 305719; mouse IgG1), T-bet-PerCP (Clone 525803; mouse IgG1), CD4-PE (Clone GK1.5; rat IgG2B)
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.