Learn More
Raybiotech Inc Quantibody Mouse Th17 Array 1 Kit.
Supplier:  Raybiotech Inc QAMTH1712
Encompass_Preferred
Detects 18 Mouse Th1, Th2, and Th17 Cytokines. Suitable for all liquid sample types. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: IFN-gamma, IL-1 beta (IL-1 F2), IL-10, IL-12 p70, IL-13, IL-17A, IL-17F, IL-2, IL-21, IL-22, IL-23 p19, IL-28A (IFN-lambda 2), IL-4, IL-5, IL-6, MIP-3 alpha (CCL20), TGF beta 1, TNF alpha
Catalog No. 50-195-0896