Learn More
BIOHIPPO Mouse THBS4 Microsample
SDP
Catalog No. 5000315638
Encompass
Mouse THBS4 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of THBS4 in plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 31 25-2000 ng/mL with a sensitivity of 9 21 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse THBS4 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse THBS4 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse THBS4 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15638 Supplier BIOHIPPO Supplier No. BHE15212503
Mouse THBS4 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of THBS4 in plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 31 25-2000 ng/mL with a sensitivity of 9 21 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse THBS4 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse THBS4 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse THBS4 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color