Learn More
Biomatik Corporation Mouse Thioredoxin Reductase 1 (TXNRD1) ELISA Kit
Supplier:  Biomatik Corporation EKU07620
Encompass_Preferred
Mouse Thioredoxin Reductase 1 (TXNRD1) ELISA Kit for detecting TXNRD1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9JMH6. Gene ID: 50493. Target Synonyms: TrxR1; TR1; GRIM12; KDRF; TRXR-1; TXNR; Gene associated with retinoic and interferon-induced mortality 12 protein; KM-102-derived reductase-like factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7095