Learn More
Sino Biological Anti-Thimet Oligopeptidase/THOP1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50828RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse THOP1 (rM THOP1; Catalog#50828-M07B; NP_073144.3; Lys 2-Cys687). THOP1 specific IgG was purified by Mouse THOP1 affinity chromatography.
Catalog No. 50-255-3063