Learn More
BIOHIPPO Mouse THP Microsample
SDP
Catalog No. 5000315612
Encompass
Mouse THP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of THP in serum plasma urine and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 057 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse THP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse THP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse THP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15612 Supplier BIOHIPPO Supplier No. BHE15211171
Mouse THP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of THP in serum plasma urine and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 057 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse THP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse THP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse THP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color