Learn More
Sino Biological Recombinant Anti-TPO/Thrombopoietin Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50146R444100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Thrombopoietin / THPO (rM Thrombopoietin / THPO; Catalog#50146-M08H; NP_033405.1; Met 1-Thr 356).
Catalog No. 50-241-2582