Learn More
Sino Biological Anti-TPO/Thrombopoietin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50146RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse THPO (rM THPO; Catalog#50146-M08H; NP_033405.1; Met 1-Thr 356). THPO specific IgG was purified by mouse THPO affinity chromatography.
Catalog No. 50-240-8962