Learn More
Sino Biological Anti-TPO/Thrombopoietin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50146T32100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Thrombopoietin/THPO (rM Thrombopoietin/THPO; Catalog#50146-M08H; NP_033405.1; Met1-Thr356). Thrombopoietin/THPO specific IgG was purified by Mouse Thrombopoietin/THPO affinity chromatography.
Catalog No. 50-240-8964