Learn More
American Research Products Inc Mouse thrombin antithrombin co
Supplier:  American Research Products Inc CSBE08433M
Encompass
Mouse thrombin-antithrombin compleX TAT ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29712