Learn More
Innovative Research Inc Mouse Thrombin Purified 50ug
Supplier:  Innovative Research Inc IMSTHR50UG
Encompass_Preferred
Mouse Thrombin Purified from Innovative Research has been purified from Mouse Plasma using activation with coastal taipan venom followed by affinity chromatography. This is a frozen liquid buffered in 0.025M Tris-HCl; 0.6M NaCl; pH 7.5, with a concentration of 0.5 mg/ml and a purity of >95%.
Catalog No. 50-203-6453