Learn More
Biomatik Corporation Mouse Thrombomodulin (TM) CLIA Kit
Supplier:  Biomatik Corporation EKU09916
Encompass_Preferred
Mouse Thrombomodulin (TM) CLIA Kit for detecting TM in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.55-4,000pg/ml. Sensitivity: 1.97pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P15306. Gene ID: 21824. Target Synonyms: CD141; THBD; BDCA3; THRM; Fetomodulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5874