Learn More
American Research Products Inc Mouse thrombospondin 1 TSP 1
Catalog No. 5000326769
Encompass
Mouse thrombospondin 1 TSP-1 ELISA Kit from CUSABIO Detection Range 3 12 ng/ml-200 ng/ml Sensitivity 1 56 ng/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26769 Supplier American Research Products Inc Supplier No. CSBE08765M
Mouse thrombospondin 1 TSP-1 ELISA Kit from CUSABIO Detection Range 3 12 ng/ml-200 ng/ml Sensitivity 1 56 ng/ml Sample serum plasma tissue homogenates Method Sandwich