Learn More
American Research Products Inc Mouse Thrombospondin 2 THBS2
Catalog No. 5000327797
Encompass
Mouse Thrombospondin-2 (THBS2) ELISA kit from CUSABIO Detection Range 6 25 ng/ml-400 ng/ml Sensitivity 1 56 ng/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27797 Supplier American Research Products Inc Supplier No. CSBEL023488MO
Mouse Thrombospondin-2 (THBS2) ELISA kit from CUSABIO Detection Range 6 25 ng/ml-400 ng/ml Sensitivity 1 56 ng/ml Sample serum plasma tissue homogenates Method Sandwich