Learn More
Sino Biological Mouse THY1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 110161T32100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Thy1 / CD90 (rM Thy1 / CD90; Catalog#50461-M08H; NP_033408.1; Met1-Cys131). Thy1 / CD90 specific IgG was purified by Mouse Thy1 / CD90 affinity chromatography.
Catalog No. 50-237-4134