Learn More
Biomatik Corporation Mouse Thymus Activation Regulated Chemokine (TARC) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11256
Encompass_Preferred
Mouse Thymus Activation Regulated Chemokine (TARC) Mini sample ELISA Kit for detecting TARC in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 10.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WUZ6. Gene ID: 20295. Target Synonyms: CCL17; ABCD-2; SCYA17; TARC; Small Inducible Cytokine Subfamily A (Cys-Cys) Member 17; Small-inducible cytokine A17. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8181