Learn More
Biomatik Corporation Mouse Thyroid Hormone Receptor Interactor 6 (TRIP6) ELISA Kit
Supplier:  Biomatik Corporation EKU07677
Encompass_Preferred
Mouse Thyroid Hormone Receptor Interactor 6 (TRIP6) ELISA Kit for detecting TRIP6 in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.144ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z1Y4. Gene ID: 22051. Target Synonyms: OIP1; ZRP-1; Opa-interacting protein 1; Zyxin-related protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8737